Titanium ≥99.99%, cubes 6.4 mm square

Supplier: BeanTown Chemical

125500-1SLUG 125500-25SLUG 125500-5SLUG
BT125500-1SLUGEA 30.59 USD
BT125500-1SLUG BT125500-25SLU BT125500-5SLUG
Titanium ≥99.99%, cubes 6.4 mm square
Titanium

CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5


  • Solid

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR