Rhodium, Plasma Standard Solution, Rh 1000 µg/ml

Supplier: BeanTown Chemical

125490-500ML
BT125490-100MLEA 495.59 USD
BT125490-100ML BT125490-500ML BT125490-50ML
Rhodium, Plasma Standard Solution, Rh 1000 µg/ml
Standards Cationic Standards Rhodium Standards

EC No: 231-595-7
UN number: UN1789
Haz class: 8
Packing group: II
Signal word: Danger

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR