4-Iodo-3-nitrobenzoic acid ≥97%

Supplier: BeanTown Chemical

125420-25G 125420-5G
BT125420-25GEA 721 USD
BT125420-25G BT125420-5G
4-Iodo-3-nitrobenzoic acid ≥97%
4-Iodo-3-nitrobenzoic acid

CAS: 35674-27-2; MDL No: MFCD00502399 Crystalline/Powder; Molecular Formula: C7H4INO4; MW: 293.02 Light Sensitive


  • Light Sensitive
  • Crystalline/Powder

Formula: C₇H₄INO₄
MDL Number: MFCD00502399
CAS Number: 35674-27-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR