3,5-Dichlorophenol ≥98%
Supplier: BeanTown Chemical
125370-25G
125370-50G
125370-5G
BT125370-25GEA
204
USD
BT125370-25G
BT125370-50G
BT125370-5G
3,5-Dichlorophenol ≥98%
3,5-Dichlorophenol
CAS: 591-35-5; EC No: 209-714-9; MDL No: MFCD00002259; RTECS: SK8820000
UN No: UN2020; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Molecular Formula: C6H4Cl2O; MW: 163.00
Melting Point: 65-68°; Boiling Point: 233°
- RTECS: SK8820000
- Crystalline/Powder
Formula:
C₆H₄Cl₂O Boiling Pt: 233 °C Melting Pt: 65…68 °C |
MDL Number:
MFCD00002259 CAS Number: 591-35-5 EINECS: 209-714-9 UN: 2020 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...