6-Chloro-m-toluidine ≥98%

Supplier: BeanTown Chemical

125335-25G 125335-5G
BT125335-25GEA 321 USD
BT125335-25G BT125335-5G
6-Chloro-m-toluidine ≥98%
6-Chloro-m-toluidine

CAS: 95-81-8; EC No: 202-454-7; MDL No: MFCD00007674 UN No: UN2239; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H8ClN ; MW: 141.60 Melting Point: 29-30°; Boiling Point: 228-230°; Flash point: 107°C (225°F)


  • Solid

Formula: C₇H₈ClN
Boiling Pt: 228…230 °C
Melting Pt: 29…30 °C
Flash Pt: 107 °C (225 °F)
MDL Number: MFCD00007674
CAS Number: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR