Bis(pinacolato)diboron ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

125265-1G 125265-25G 125265-5G
BT125265-1GEA 39 USD
BT125265-1G BT125265-25G BT125265-5G
Bis(pinacolato)diboron ≥98%
Bis(pinacolato)diboron

CAS: 73183-34-3; MDL No: MFCD00799570 Powder; Molecular Formula: C12H24B2O4; MW: 253.94 Melting Point: 137-140°


  • Powder

Formula: C₁₂H₂₄B₂O₄
MW: 253.94 g/mol
Melting Pt: 137…140 °C
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR