Bis(pinacolato)diboron ≥98%
Supplier: BeanTown Chemical
Synonyms:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
125265-1G
125265-25G
125265-5G
BT125265-1GEA
39
USD
BT125265-1G
BT125265-25G
BT125265-5G
Bis(pinacolato)diboron ≥98%
Bis(pinacolato)diboron
CAS: 73183-34-3; MDL No: MFCD00799570
Powder; Molecular Formula: C12H24B2O4; MW: 253.94
Melting Point: 137-140°
- Powder
Formula:
C₁₂H₂₄B₂O₄ Melting Pt: 137…140 °C |
MDL Number:
MFCD00799570 CAS Number: 73183-34-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...