Indium (III) oxide ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Indium trioxide, Diindium trioxide

125190-10G 125190-50G
BT125190-10GEA 95.85 USD
BT125190-10G BT125190-50G
Indium (III) oxide ≥99.999%
Indium (III) oxide

CAS: 1312-43-2; EC No: 215-193-9; MDL No: MFCD00011060 3-6mm Sintered Granules; Molecular Formula: In2O3; MW: 277.64 Density (g/mL): 7.18


  • 3-6mm Sintered Granules

Formula: In₂O₃
Density: 7.18 g/mL
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR