Bismuth(III) bromide ≥99%

Supplier: BeanTown Chemical

125160-100G 125160-25G
BT125160-100GEA 324 USD
BT125160-100G BT125160-25G
Bismuth(III) bromide ≥99%
Bismuth (III) bromide

CAS: 7787-58-8; EC No: 232-121-1; MDL No: MFCD00014195 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Molecular Formula: BiBr3; MW: 448.69 Density (g/mL): 5.7 Hygroscopic


  • Hygroscopic
  • Powder

Formula: BiBr₃
MW: 448.69 g/mol
Boiling Pt: 278
Melting Pt: 218
Density: 5.7 g/mL
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8 III
Merck Index: 12,01299

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR