Gallium antimonide ≥99.99%

Supplier: BeanTown Chemical

125065-1G 125065-5G
BT125065-1GEA 53 USD
BT125065-1G BT125065-5G
Gallium antimonide ≥99.99%
Gallium antimonide

CAS: 12064-03-8; EC No: 235-058-8; MDL No: MFCD00016101 UN No: UN1549; Haz Class: 6.1; Packing Group: III 6mm & Down Pieces; Molecular Formula: GaSb; MW: 191.47 Melting Point: 710° Density (g/mL): 5.619


  • 6mm & Down Pieces

Formula: GaSb
Melting Pt: 710 °C
Density: 5.619 g/mL
MDL Number: MFCD00016101
CAS Number: 12064-03-8
EINECS: 235-058-8
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR