Titanium diboride ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Titanium boride

125010-10G 125010-250G 125010-50G
BT125010-10GEA 40 USD
BT125010-10G BT125010-250G BT125010-50G
Titanium diboride ≥99.5%
Titanium diboride

CAS: 12045-63-5; EC No: 234-961-4; MDL No: MFCD00049595 -325 Mesh Powder; Linear Formula: TiB2; MW: 69.50 Melting Point: 3225° Density (g/mL): 4.52


  • -325 Mesh Powder

Formula: B₂Ti
Melting Pt: 3225 °C
Density: 4.52 g/mL
MDL Number: MFCD00049595
CAS Number: 12045-63-5
EINECS: 234-961-4

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR