Ammonium carbonate ≥30% (NH₃ basis) ACS
Supplier: BeanTown Chemical
125005-2KG
125005-500G
BT125005-2KGEA
133
USD
BT125005-2KG
BT125005-500G
Ammonium carbonate ≥30% (NH₃ basis) ACS
Ammonium carbonate
CAS: 506-87-6; EC No: 208-058-0; MDL No: MFCD00010890; RTECS: BP1925000
Powder; Linear Formula: (NH4)2CO3; MW: 96.09
Hygroscopic
- RTECS: BP1925000
- Hygroscopic
- Powder
Formula:
(NH₄)₂CO₃ |
MDL Number:
MFCD00010890 CAS Number: 506-87-6 EINECS: 208-058-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...