Glycylglycine (Gly-Gly)

Supplier: BeanTown Chemical

124900-100G 124900-25G 124900-500G
BT124900-100GEA 114 USD
BT124900-100G BT124900-25G BT124900-500G
Glycylglycine (Gly-Gly)
Proteins and Peptides

EINECS: 209-127-8
Melting point: 225 °C (dec.)
Density at 20 °C (68 °F): 0.35 g/cm³ (2.92075 lbs/gal)
Signal word: Warning
GHS symbol: GHS07
Merck number: 14,4503


Delivery information: Non-hazardous for shipping.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR