Ammonium molybdate (para) hydrate ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Ammonium molybdate hydrate
124875-25G
124875-5G
BT124875-25GEA
170.08
USD
BT124875-25G
BT124875-5G
Ammonium molybdate (para) hydrate ≥99.999%
Ammonium molybdate (para) hydrate
CAS: 12027-67-7; EC No: 234-722-4; MDL No: MFCD00064633; RTECS: QA4900000
Powder; Linear Formula: (NH4)6Mo7O24·H2O; MW: 1163.79 (anhydrous)
Density (g/mL): 2.498
- RTECS: QA4900000
- Powder
Formula:
H₂₄Mo₇N₆O₂₄·xH₂O Density: 2.498 g/mL |
MDL Number:
MFCD00064633 CAS Number: 12027-67-7 EINECS: 234-722-4 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...