Calcium metasilicate ≥99%

Supplier: BeanTown Chemical

Synonyms: Calcium silicate

124870-100G 124870-25G 124870-500G
BT124870-100GEA 164 USD
BT124870-100G BT124870-25G BT124870-500G
Calcium metasilicate ≥99%
Calcium metasilicate

CAS: 10101-39-0; EC No: 233-250-6; MDL No: MFCD00015979 -200 Mesh Powder; Linear Formula: CaSiO3 ; MW: 116.16


  • -200 Mesh Powder

Formula: CaO₃Si
MW: 116.16 g/mol
MDL Number: MFCD00015979
CAS Number: 10101-39-0
EINECS: 233-250-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR