Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
124850-1G
124850-25G
124850-5G
BT124850-1GEA
50
USD
BT124850-1G
BT124850-25G
BT124850-5G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99.9%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II)
CAS: 14040-05-2; MDL No: MFCD00058920
Powder; Molecular Formula: C22H38CuO4; MW: 430.08
Melting Point: 198° (decomposes)
- Powder
Formula:
C₂₂H₃₈CuO₄ Melting Pt: 198 °C (dec.) |
MDL Number:
MFCD00058920 CAS Number: 14040-05-2 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...