Potassium carbonate ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt
124785-100G
124785-25G
124785-5G
BT124785-100GEA
264
USD
BT124785-100G
BT124785-25G
BT124785-5G
Potassium carbonate ≥99.995%
Potassium carbonate
CAS: 584-08-7; EC No: 209-529-3; MDL No: MFCD00011382; RTECS: TS7750000
Powder; Linear Formula: K2CO3; MW: 138.21
Melting Point: 891°
Density (g/mL): 2.43
Hygroscopic
- RTECS: TS7750000
- Hygroscopic
- Powder
Formula:
K₂CO₃ MW: 138.21 g/mol Melting Pt: 891 Density: 2.43 g/mL |
MDL Number:
MFCD00011382 CAS Number: 584-08-7 EINECS: 209-529-3 Merck Index: 13,07702 |
|
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...