2,2',5,5'-Tetrachlorobenzidine ≥98%
Supplier: BeanTown Chemical
Synonyms:
4,4'-Diamino-2,2',5,5'-tetrachlorobiphenyl, 2,2,5,5-Tetrachlorobenzidine
124770-25G
BT124770-25GEA
65
USD
BT124770-25G
2,2',5,5'-Tetrachlorobenzidine ≥98%
2,2',5,5'-Tetrachlorobenzidine
CAS: 15721-02-5; EC No: 239-815-3; MDL No: MFCD00043918; RTECS: DD1751000
Solid; Molecular Formula: C12H8Cl4N2; MW: 322.02
Melting Point: 135-140°; Boiling Point: 408°
- RTECS: DD1751000
- Solid
Formula:
C₁₂H₈Cl₄N₂ Boiling Pt: 408 °C Melting Pt: 135…140 °C |
MDL Number:
MFCD00043918 CAS Number: 15721-02-5 EINECS: 239-815-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...