4-(Trifluoromethoxy)phenyl isocyanate ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

124765-1G 124765-25G 124765-5G
BT124765-1GEA 54 USD
BT124765-1G BT124765-25G BT124765-5G
4-(Trifluoromethoxy)phenyl isocyanate ≥97%
4-(Trifluoromethoxy)phenyl isocyanate

CAS: 35037-73-1; EC No: 252-328-0; MDL No: MFCD00035702 UN No: UN2206; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C8H4F3NO2; MW: 203.12 Boiling Point: 78°/22 mmHg; Flash point: 69°C (156°F) Density (g/mL): 1.346; Refractive Index: 1.458 Moisture Sensitive


  • Refractive index: 1.458
  • Moisture Sensitive
  • Liquid

Formula: C₈H₄F₃NO₂
Boiling Pt: 78 °C/22 mmHg
Density: 1.346 g/mL
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00035702
CAS Number: 35037-73-1
EINECS: 252-328-0
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR