Lead(IV) oxide ≥97.0% ACS

Supplier: BeanTown Chemical

124725-100G 124725-2KG 124725-500G
BT124725-100GEA 68.76 USD
BT124725-100G BT124725-2KG BT124725-500G
Lead(IV) oxide ≥97.0% ACS
Lead (IV) oxide

CAS: 1309-60-0; EC No: 215-174-5; MDL No: MFCD00011165 UN No: UN1872; Haz Class: 5.1; Packing Group: III Powder; Linear Formula: PbO2; MW: 239.19


  • Powder

Formula: O₂Pb
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1,III

Order Now

Specification Test Results

Assay 97.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR