Iron(II) acetate, anhydrous

Supplier: BeanTown Chemical

124680-100G 124680-25G
BT124680-100GEA 584.71 USD
BT124680-100G BT124680-25G
Iron(II) acetate, anhydrous
Iron (II) acetate

CAS: 3094-87-9; EC No: 221-441-7; MDL No: MFCD00058909 Powder/Lump; Linear Formula: Fe(OOCCH3)2; MW: 173.94 Melting Point: 199-200° (decomposes) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder/Lump

Formula: C₄H₆FeO₄
Melting Pt: 199…200 °C (dec.)
MDL Number: MFCD00058909
CAS Number: 3094-87-9
EINECS: 221-441-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR