1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,1,1,3,3,3-Hexafluoro-2-phenylpropan-2-ol, alpha,alpha-Bis(trifluoromethyl)benzyl Alcohol, 2,2,2,2',2',2'-Hexafluorocumyl Alcohol

124650-1G 124650-25G 124650-5G
BT124650-1GEA 20 USD
BT124650-1G BT124650-25G BT124650-5G
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol ≥98%
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol

CAS: 718-64-9; EC No: 211-943-4; MDL No: MFCD00040983 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C9H6F6O; MW: 244.13 Boiling Point: 160°; Flash point: 60°C (140°F) Density (g/mL): 1.45; Refractive Index: 1.415


  • Refractive index: 1.415
  • Liquid

Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 160 °C
Density: 1.45 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00040983
CAS Number: 718-64-9
EINECS: 211-943-4
ADR: 3 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR