β-(N-Morpholino)ethanesulfonic acid (MES) hydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
4-Morpholineethanesulphonic acid hydrate, MES hydrate, 2-(4-Morpholinyl)ethanesulphonic acid hydrate, 4-Morpholineethanesulfonic acid hydrate, 2-(4-Morpholinyl)ethanesulfonic acid hydrate
124645-250G
124645-500G
BT124645-250GEA
203.74
USD
BT124645-250G
BT124645-500G
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate ≥99%
β-(N-Morpholino)ethanesulfonic acid (MES) hydrate
CAS: 4432-31-9; EC No: 224-632-3; MDL No: MFCD00149409; RTECS: QE9104530
Powder; Molecular Formula: C6H13NO4S·xH2O; MW: 195.24
Melting Point: 308° (decomposes)
Formula:
C₆H₁₃NO₄S·xH₂O |
MDL Number:
MFCD00149409 CAS Number: 1266615-59-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...