6-Bromopicolinic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
124555-1G
124555-5G
BT124555-1GEA
53
USD
BT124555-1G
BT124555-5G
6-Bromopicolinic acid ≥97%
6-Bromopicolinic acid
CAS: 21190-87-4; MDL No: MFCD00093197
Crystalline; Molecular Formula: C6H3BrNO2; MW: 202.01
Melting Point: 192-194°
- Crystalline
Formula:
C₆H₄BrNO₂ Melting Pt: 192…194 °C |
MDL Number:
MFCD00093197 CAS Number: 21190-87-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...