Yttrium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.64 mm
Supplier: BeanTown Chemical
124500-25X25MM
124500-50X50MM
BT124500-25X25EA
146
USD
BT124500-25X25
BT124500-50X50
Yttrium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.64 mm
Yttrium
CAS: 7440-65-5; EC No: 231-174-8; MDL No: MFCD00011468
Solid; Molecular Formula: Y; MW: 88.91
Melting Point: 1522°; Boiling Point: 3338°
Density (g/mL): 4.469
- Solid
Formula:
Y Boiling Pt: 3338 °C Melting Pt: 1522 °C Density: 4.469 g/mL |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...