Soda lime ACS, with indicator

Supplier: BeanTown Chemical

124385-2.5KG 124385-500G
BT124385-2.5KGEA 351.84 USD
BT124385-2.5KG BT124385-500G
Soda lime ACS, with indicator
Soda lime

CAS: 8006-28-8; MDL No: MFCD00134094 UN No: UN1907; Haz Class: 8; Packing Group: III 4-8 Mesh Solid; Molecular Formula: CaO/NaOH Air Sensitive , Hygroscopic


  • Air Sensitive, Hygroscopic
  • 4-8 Mesh Solid

MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8 III
Merck Index: 13,08640

Order Now

Specification Test Results

Loss on drying at 200°C ≤7%
Carbon dioxide absorption capacity ≥19%
Fines 1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR