Aluminium oxide, γ-α-phase ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

124345-100G 124345-25G 124345-500G
BT124345-100GEA 116 USD
BT124345-100G BT124345-25G BT124345-500G
Aluminium oxide, γ-α-phase ≥99.99% (trace metals basis)
Aluminium oxide, γ-α-phase

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 0.3 Micron Powder, S.A. 15 m2/g; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 0.3 Micron Powder, S.A. 15 m²/g

Formula: Al₂O₃
MW: 101.96
Boiling Pt: 2980
Melting Pt: 2050
Density: 3.97
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 12,00369

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR