(±)-1-Octen-3-ol ≥98%

Supplier: BeanTown Chemical

Synonyms: (±)-Oct-1-en-3-ol, (±)-Pentyl vinyl carbinol, Oct-1-en-3-ol

124335-10G 124335-250G 124335-50G
BT124335-10GEA 32 USD
BT124335-10G BT124335-250G BT124335-50G
(±)-1-Octen-3-ol ≥98%
(±)-1-Octen-3-ol

CAS: 3391-86-4; EC No: 222-226-0; MDL No: MFCD00004589; RTECS: RH3300000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: CH3(CH2)4CH(OH)CH=CH2; Molecular Formula: C8H16O; MW: 128.21 Boiling Point: 84-85°/25 mmHg; Flash point: 68°C (154°F) Density (g/mL): 0.83; Refractive Index: 1.437


  • RTECS: RH3300000
  • Refractive index: 1.437
  • Liquid

Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 174
Melting Pt: -49
Density: 0.83 g/mL
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD00004589
CAS Number: 3391-86-4
EINECS: 222-226-0
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=852e188ec9a8f0b95fb8267eecb509cc8b9b4817&vid=db554820fe02a5820107eb97e322b46f3310f665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR