Iron(II) sulfide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Ferrous sulphide, Ferrous sulfide

124300-10G 124300-50G
BT124300-10GEA 55 USD
BT124300-10G BT124300-50G
Iron(II) sulfide ≥99.9%
Iron (II) sulphide

CAS: 1317-37-9; EC No: 215-268-6; MDL No: MFCD00011013 -100 Mesh Powder; Molecular Formula: FeS; MW: 87.91 Density (g/mL): 4.74 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • -100 Mesh Powder

Formula: FeS
Density: 4.74 g/mL
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR