Calcium chloride, anhydrous ≥93%, porous
Supplier: BeanTown Chemical
124285-2KG
124285-500G
BT124285-2KGEA
122
USD
BT124285-2KG
BT124285-500G
Calcium chloride, anhydrous ≥93%, porous
Calcium chloride
CAS: 10043-52-4; EC No: 233-140-8; MDL No: MFCD00010903; RTECS: EV9800000
-4 Mesh Powder; Molecular Formula: CaCl2 ; MW: 110.98
Melting Point: 772°
Hygroscopic
- RTECS: EV9800000
- Hygroscopic
- -4 Mesh Powder
Formula:
CaCl₂ Melting Pt: 772 °C |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 |
Specification Test Results
Assay | 93% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...