Zinc chloride ≥97.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
124275-1KG
124275-250G
BT124275-1KGEA
173.54
USD
BT124275-1KG
BT124275-250G
Zinc chloride ≥97.0% ACS
Zinc chloride
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000
UN No: UN2331; Haz Class: 8; Packing Group: III
Powder; Linear Formula: ZnCl2; MW: 136.29
Melting Point: 293°; Boiling Point: 732°
Density (g/mL): 2.907
Hygroscopic
- RTECS: ZH1400000
- Hygroscopic
- Powder
Formula:
ZnCl₂ Boiling Pt: 732 °C Melting Pt: 293 °C Density: 2.907 g/mL |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III |
Specification Test Results
Assay | 97.0% min. |
Oxycloride | Passes test |
Insoluble matter | ≤0.005% |
Nitrate (NO3) | ≤0.003% |
Sulfate (SO4) | ≤0.01% |
Ammonium (NH4) | ≤0.005% |
Calcium (Ca) | ≤0.06% |
Iron (Fe) | ≤0.001% |
Lead (Pb) | ≤0.005% |
Magnesium (Mg) | ≤0.01% |
Potassium (K) | ≤0.02% |
Sodium (Na) | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...