Zinc chloride ≥97.0% ACS

Supplier: BeanTown Chemical

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

124275-1KG 124275-250G
BT124275-1KGEA 173.54 USD
BT124275-1KG BT124275-250G
Zinc chloride ≥97.0% ACS
Zinc chloride

CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000 UN No: UN2331; Haz Class: 8; Packing Group: III Powder; Linear Formula: ZnCl2; MW: 136.29 Melting Point: 293°; Boiling Point: 732° Density (g/mL): 2.907 Hygroscopic


  • RTECS: ZH1400000
  • Hygroscopic
  • Powder

Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 293 °C
Density: 2.907 g/mL
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III

Order Now

Specification Test Results

Assay 97.0% min.
Oxycloride Passes test
Insoluble matter ≤0.005%
Nitrate (NO3) ≤0.003%
Sulfate (SO4) ≤0.01%
Ammonium (NH4) ≤0.005%
Calcium (Ca) ≤0.06%
Iron (Fe) ≤0.001%
Lead (Pb) ≤0.005%
Magnesium (Mg) ≤0.01%
Potassium (K) ≤0.02%
Sodium (Na) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR