Methanol absolute ≥99.8% ACS low acid

Supplier: BeanTown Chemical
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

124220-1L 124220-4L 124220-500ML
BT124220-1LEA 29 USD
BT124220-1L BT124220-4L BT124220-500ML
Methanol absolute ≥99.8% ACS low acid
Methanol

CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3OH; MW: 32.04 Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F) Density (g/mL): 0.791; Refractive Index: 1.329


  • RTECS: PC1400000
  • Refractive index: 1.329
  • Liquid

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.791 g/mL
Flash Pt: 9.7 °C (49.5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8% min.
Color (APHA) ≤10
Residue after evaporation ≤0.001%
Identification, IR Passes test
Identification, GC Passes test
Acidity ≤0.45ml, 0.02N NaOH
Alkalinity (as ammonia) ≤0.20ml, 0.02N H2SO4
Titrable acid ≤0.0003meq/g
Titrable base ≤0.0002meq/g
Readily carbonizable substances Passes test
Acetone & aldehydes ≤0.003%
Readily oxidizable substances Passes test
Non-volatile residue ≤2mg (0.001% w/w)
Water ≤0.10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR