1-Butanesulfonyl chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Butanesulphonylchloride, 1-Butanesulfonylchloride
124115-100G
124115-25G
124115-5G
BT124115-100GEA
253
USD
BT124115-100G
BT124115-25G
BT124115-5G
1-Butanesulfonyl chloride ≥98%
1-Butanesulphonyl chloride
CAS: 2386-60-9; EC No: 219-204-8; MDL No: MFCD00007464; RTECS: EK5950000
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: CH3(CH2)3SO2Cl; Molecular Formula: C4H9ClO2S; MW: 156.63
Boiling Point: 80-81°/9 mmHg; Flash point: 79°C (174°F)
Density (g/mL): 1.208; Refractive Index: 1.454
Moisture Sensitive
- RTECS: EK5950000
- Refractive index: 1.454
- Moisture Sensitive
- Liquid
Formula:
C₄H₉ClO₂S MW: 156.63 g/mol Boiling Pt: 208 Melting Pt: -25 Density: 1.208 g/mL Flash Pt: 79 °C (174 °F) |
MDL Number:
MFCD00007464 CAS Number: 2386-60-9 EINECS: 219-204-8 UN: 3265 ADR: 8 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...