Rhodium(III) chloride, anhydrous ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Rhodium(III) chloride, Rhodium trichloride

124075-1G 124075-250MG
BT124075-1GEA 605.55 USD
BT124075-1G BT124075-250MG
Rhodium(III) chloride, anhydrous ≥99.9%
Rhodium (III) chloride

CAS: 10049-07-7; EC No: 233-165-4; MDL No: MFCD00011204; RTECS: VI9275000 Crystalline/Powder; Linear Formula: RhCl3; MW: 209.26 Hygroscopic


  • RTECS: VI9275000
  • Hygroscopic
  • Crystalline/Powder

Formula: Cl₃Rh
MW: 209.26
Density: 5.38
MDL Number: MFCD00011204
CAS Number: 10049-07-7
EINECS: 233-165-4
Merck Index: 12,08355

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR