Mercury(II) thiocyanate
Supplier: BeanTown Chemical
CAS: 592-85-8; EC No: 209-773-0; MDL No: MFCD00011039; RTECS: XL1550000
UN No: UN1646; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: Hg(SCN)2; MW: 316.75
Melting Point: 165° (decomposes); Flash point: 120°C (248°F)
Density (g/mL): 3.71
Light Sensitive, Moisture Sensitive
- RTECS: XL1550000
- Light Sensitive, Moisture Sensitive
- Powder
Formula:
C₂HgN₂S₂ Melting Pt: 165 °C (dec.) Density: 3.71 g/mL Flash Pt: 120 °C (248 °F) |
MDL Number:
MFCD00011039 CAS Number: 592-85-8 EINECS: 209-773-0 UN: 1646 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...