Acetone-[D₆] ≥99.9% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

123980-10G 123980-5X10G
BT123980-10GEA 104.63 USD
BT123980-10G BT123980-5X10G
Acetone-[D₆] ≥99.9% (Isotopic)
Acetone-[D6]

CAS: 666-52-4; EC No: 211-563-9; MDL No: MFCD00044635 UN No: UN1090; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CD3COCD3; MW: 64.12 Melting Point: -94°; Boiling Point: 55.5°; Flash point: -19°C (-2°F) Density (g/mL): 0.872; Refractive Index: 1.355


  • Refractive index: 1.355
  • Liquid

Formula: D₃CC(O)CD₃
Boiling Pt: 55.5 °C
Melting Pt: –94 °C
Density: 0.872 g/mL
Flash Pt: –19 °C (–2 °F)
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR