Potassium nitrate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Nitrate of Potash

123945-100G 123945-2KG 123945-500G
BT123945-100GEA 44.58 USD
BT123945-100G BT123945-2KG BT123945-500G
Potassium nitrate ≥99.0% ACS
Potassium nitrate

CAS: 7757-79-1; EC No: 231-818-8; MDL No: MFCD00011409; RTECS: TT3700000 UN No: UN1486; Haz Class: 5.1; Packing Group: III Crystalline; Molecular Formula: KNO3; MW: 101.11 Melting Point: 334°; Boiling Point: 400° (decomposes) Density (g/mL): 2.109 Hygroscopic


  • RTECS: TT3700000
  • Hygroscopic
  • Crystalline

Formula: KNO₃
MW: 101.1 g/mol
Boiling Pt: 400 °C (dec.)
Melting Pt: 334
Density: 2.109 g/mL
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1 III
Merck Index: 13,07733

Order Now

Specification Test Results

Assay 99.0% min.
pH of 5% solution at 25°C 4.5-8.5
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.002%
Iodate (IO3) ≤5ppm
Nitrite (NO2) ≤0.001%
Phosphate (PO4) ≤5ppm
Sulfate (SO4) ≤0.003%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤3ppm
Calcium (Ca) ≤0.005%
Magnesium (Mg) ≤0.002%
Sodium (Na) ≤0.005%
Solubility 10% solution in water is clear and colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR