Allyltrimethylsilane ≥98%

Supplier: BeanTown Chemical

123875-10G 123875-250G 123875-50G
BT123875-10GEA 45.2 USD
BT123875-10G BT123875-250G BT123875-50G
Allyltrimethylsilane ≥98%
Allyltrimethylsilane

CAS: 762-72-1; EC No: 212-104-5; MDL No: MFCD00008635 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH2=CHCH2Si(CH3)3; Molecular Formula: C6H14Si; MW: 114.26 Boiling Point: 84-88°; Flash point: 16°C (61°F) Density (g/mL): 0.719; Refractive Index: 1.407


  • Refractive index: 1.407
  • Liquid

Formula: H₂CCHCH₂Si(CH₃)₃
Boiling Pt: 84…88 °C
Density: 0.719 g/mL
Flash Pt: 16 °C (61 °F)
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR