4-Aminobenzyl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol

123865-25G 123865-5G
BT123865-25GEA 388 USD
BT123865-25G BT123865-5G
4-Aminobenzyl alcohol ≥98%
4-Aminobenzyl alcohol

CAS: 623-04-1; EC No: 210-767-5; MDL No: MFCD00014782 Crystalline; Molecular Formula: C7H9NO; MW: 123.15 Melting Point: 60-65° Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₇H₉NO
MW: 123.15
Melting Pt: 75
MDL Number: MFCD00014782
CAS Number: 623-04-1
EINECS: 210-767-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR