Nitrobenzene ≥99%

Supplier: BeanTown Chemical

123770-10KG 123770-2.5KG 123770-500G
BT123770-10KGEA 244 USD
BT123770-10KG BT123770-2.5KG BT123770-500G
Nitrobenzene ≥99%
Nitrobenzene

CAS: 98-95-3; EC No: 202-716-0; MDL No: MFCD00007043; RTECS: DA6475000 UN No: UN1662; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C6H5NO2; MW: 123.11 Melting Point: 5-6°; Boiling Point: 210-211°; Flash point: 87°C (188°F) Density (g/mL): 1.196; Refractive Index: 1.551


  • RTECS: DA6475000
  • Refractive index: 1.551
  • Liquid

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210.8
Melting Pt: 5.7
Density: 1.196 g/mL
Flash Pt: 87 °C (188 °F)
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1 II
Merck Index: 13,06621

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR