Cesium acetate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetic acid cesium salt
123715-250G
123715-50G
BT123715-250GEA
213.95
USD
BT123715-250G
BT123715-50G
Cesium acetate ≥99%
Cesium acetate
CAS: 3396-11-0; EC No: 222-248-0; MDL No: MFCD00013056
Crystalline/Powder; Linear Formula: CsOOCCH3; MW: 191.95
Melting Point: 194°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
H₃CCOOCs Melting Pt: 194 °C |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 EINECS: 222-248-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...