Potassium thiocyanate ≥98.5% ACS
Supplier: BeanTown Chemical
CAS: 333-20-0; EC No: 206-370-1; MDL No: MFCD00011413; RTECS: XL1925000
Crystalline; Linear Formula: KSCN; MW: 97.18
Melting Point: 173°; Boiling Point: 500° (decomposes)
Density (g/mL): 1.89
Light Sensitive, Moisture Sensitive
- RTECS: XL1925000
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
KSCN Boiling Pt: 500 °C (dec.) Melting Pt: 173 °C Density: 1.89 g/mL |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 EINECS: 206-370-1 |
Specification Test Results
Assay | 98.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...