Potassium thiocyanate ≥98.5% ACS

Supplier: BeanTown Chemical

123700-100G 123700-500G
BT123700-100GEA 55.65 USD
BT123700-100G BT123700-500G
Potassium thiocyanate ≥98.5% ACS
Potassium thiocyanate
Formula: KSCN
Boiling Pt: 500 °C (dec.)
Melting Pt: 173 °C
Density: 1.89 g/mL
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR