Dihydrogen hexafluorozirconate 45% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: Zirconate(2-), hexafluoro-, Hexafluorozirconic acid, Hexafluorozirconate(2-)

123645-2KG 123645-500G
BT123645-2KGEA 178 USD
BT123645-2KG BT123645-500G
Dihydrogen hexafluorozirconate 45% (w/w) in water
Dihydrogen hexafluorozirconate

CAS: 12021-95-3; MDL No: MFCD00082965 UN No: UN3264; Haz Class: 8; Packing Group: II Liquid; Linear Formula: H2ZrF6; MW: 207.22 Density (g/mL): 1.51


  • Liquid

Formula: H₂ZrF₆
Density: 1.51 g/mL
MDL Number: MFCD00082965
CAS Number: 12021-95-3
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR