3-Cyano-4-methylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile

123575-1G 123575-25G 123575-5G
BT123575-1GEA 46 USD
BT123575-1G BT123575-25G BT123575-5G
3-Cyano-4-methylpyridine ≥97%
3-Cyano-4-methylpyridine

CAS: 5444-01-9; MDL No: MFCD00234272 Crystalline; Molecular Formula: C7H6N2 ; MW: 118.14


  • Crystalline

Formula: C₇H₆N₂
MDL Number: MFCD00234272
CAS Number: 5444-01-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR