2-Iodophenol ≥98%

Supplier: BeanTown Chemical

123565-100G 123565-25G 123565-5G
BT123565-100GEA 370 USD
BT123565-100G BT123565-25G BT123565-5G
2-Iodophenol ≥98%
2-Iodophenol

CAS: 533-58-4; EC No: 208-569-9; MDL No: MFCD00013963; RTECS: SL5500000 Crystalline; Molecular Formula: C6H5IO; MW: 220.01 Melting Point: 37-40°; Boiling Point: 186-187°/160 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.947 Light Sensitive


  • RTECS: SL5500000
  • Light Sensitive
  • Crystalline

Formula: C₆H₅IO
Boiling Pt: 186…187 °C/160 mmHg
Melting Pt: 37…40 °C
Density: 1.947 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR