9-Anthracenemethanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 9-Anthrylmethanol, Anthracene-9-methanol

123530-10G 123530-250G 123530-50G
BT123530-10GEA 61.78 USD
BT123530-10G BT123530-250G BT123530-50G
9-Anthracenemethanol ≥97%
9-Anthracenemethanol

CAS: 1468-95-7; EC No: 215-998-5; MDL No: MFCD00001264; RTECS: CB0577500 Solid; Molecular Formula: C15H12O; MW: 208.26


  • RTECS: CB0577500
  • Solid

Formula: C₁₅H₁₂O
MDL Number: MFCD00001264
CAS Number: 1468-95-7
EINECS: 215-998-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR