Ammonia solution 10% (v/v) in water

Supplier: BeanTown Chemical

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

123380-1L 123380-4L 123380-500ML
BT123380-1LEA 61.61 USD
BT123380-1L BT123380-4L BT123380-500ML
Ammonia solution 10% (v/v) in water
Ammonia solution

CAS: 1336-21-6; EC No: 215-647-6; MDL No: MFCD00066650 UN No: UN2672; Haz Class: 8; Packing Group: III Liquid; Linear Formula: NH4OH; MW: 35.05 Density (g/mL): 0.9


  • Liquid

Formula: NH₃
MW: 35.05 g/mol
Density: 0.9 g/mL
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR