Tin(II) chloride, anhydrous ≥98%

Supplier: BeanTown Chemical

Synonyms: Stannous chloride, Tin dichloride

123330-100G 123330-2.5KG 123330-500G
BT123330-100GEA 52.1 USD
BT123330-100G BT123330-2.5KG BT123330-500G
Tin(II) chloride, anhydrous ≥98%
Tin (II) chloride

CAS: 7772-99-8; EC No: 231-868-0; MDL No: MFCD00011241; RTECS: XP8700000 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Linear Formula: SnCl2; MW: 189.62 Melting Point: 246°; Boiling Point: 652° Density (g/mL): 3.95 Air Sensitive, Moisture Sensitive


  • RTECS: XP8700000
  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 623
Melting Pt: 246
Density: 3.95 g/mL
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR